accessibility menu, dialog, popup

UserName

CAS RN 3528-58-3

H3CNNH2N

CAS RN 3528-58-3

IUPAC Name: 1-ethyl-1H-pyrazol-5-amine
Synonyms: 2-ethylpyrazol-3-amine
Molecular Weight (g/mol): 111.15
Molecular Formula: C5H9N3
InChi Key: FSJOLBAFVKSQQJ-UHFFFAOYSA-N
SMILES: CCN1N=CC=C1N
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results