accessibility menu, dialog, popup

UserName

CH3H3CMgCl

CAS RN 35293-35-7

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 192.97
Molecular Formula: C10H13ClMg
InChi Key: IYOKDPFKCXZKJU-UHFFFAOYSA-M
SMILES: [Mg++].[Cl-].CC(C)([CH2-])C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results