accessibility menu, dialog, popup

UserName

CAS RN 3530-15-2

CH3NNH3CNH2N

CAS RN 3530-15-2

IUPAC Name: dimethyl-4H-1,2,4-triazol-4-amine
Synonyms: dimethyl-1,2,4-triazol-4-amine
Molecular Weight (g/mol): 112.14
Molecular Formula: C4H8N4
InChi Key: MIIKMZAVLKMOFM-UHFFFAOYSA-N
SMILES: CC1=NN=C(C)N1N
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)