accessibility menu, dialog, popup

UserName

FFFFFFFFFFFFFF

CAS RN 355-02-2

IUPAC Name: 1,1,2,2,3,3,4,4,5,5,6-undecafluoro-6-(trifluoromethyl)cyclohexane
Synonyms: perfluoromethylcyclohexane
Molecular Weight (g/mol): 350.06
Molecular Formula: C7F14
InChi Key: QIROQPWSJUXOJC-UHFFFAOYSA-N
SMILES: FC(F)(F)C1(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C1(F)F
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)