accessibility menu, dialog, popup

UserName

H3COFFFFFFF

CAS RN 355-17-9

IUPAC Name: 3,3,4,4,5,5,5-heptafluoropentan-2-one
Synonyms: 3,3,4,4,5,5,5-heptafluoropentan-2-one
Molecular Weight (g/mol): 212.07
Molecular Formula: C5H3F7O
InChi Key: XJYXROGTQYHLTH-UHFFFAOYSA-N
SMILES: CC(=O)C(F)(F)C(F)(F)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results