CAS RN 356-86-5
IUPAC Name:
2,2,3,3,3-pentafluoropropyl prop-2-enoate
Synonyms:
2,2,3,3,3-pentafluoropropyl prop-2-enoate
Molecular Weight (g/mol):
204.1
Molecular Formula:
C6H5F5O2
InChi Key:
JDVGNKIUXZQTFD-UHFFFAOYSA-N
SMILES:
FC(F)(F)C(F)(F)COC(=O)C=C
1
–
1
of
1
results
1
2,2,3,3,3-Pentafluoropropyl acrylate, 97%
CAS: 356-86-5 Molecular Formula: C6H5F5O2 Molecular Weight (g/mol): 204.10 MDL Number: MFCD00039257 InChI Key: JDVGNKIUXZQTFD-UHFFFAOYSA-N Synonym: 2,2,3,3,3-pentafluoropropyl acrylate,1h,1h-pentafluoropropyl acrylate,2-propenoic acid, 2,2,3,3,3-pentafluoropropyl ester,acmc-20aokp,acrylic acid=2,2,3,3,3-pentafluoropropyl ester,acrylic acid 2,2,3,3,3-pentafluoropropyl ester,2-propenoic acid,2,2,3,3,3-pentafluoropropyl ester,2,2,3,3,3-pentafluoropropyl acrylate stabilized with tbc,2,2,3,3,3-pentafluoropropyl acrylate, contains 100 ppm 4-tert-butylcatechol as inhibitor PubChem CID: 67744 IUPAC Name: 2,2,3,3,3-pentafluoropropyl prop-2-enoate SMILES: FC(F)(F)C(F)(F)COC(=O)C=C