accessibility menu, dialog, popup

UserName

CAS RN 35700-48-2

IUPAC Name: 7-bromo-5-methyl-1-benzofuran
Synonyms: 7-bromo-5-methyl-1-benzofuran
Molecular Weight (g/mol): 211.06
Molecular Formula: C9H7BrO
InChi Key: TZUMDTURHSPEAY-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=C2OC=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results