accessibility menu, dialog, popup

UserName

CAS RN 357263-41-3

HNNCl

CAS RN 357263-41-3

IUPAC Name: 7-chloro-1H-pyrrolo[2,3-c]pyridine
Synonyms: 7-chloro-1H-pyrrolo[2,3-c]pyridine
Molecular Weight (g/mol): 152.58
Molecular Formula: C7H5ClN2
InChi Key: HOHKYYCVFMEBGG-UHFFFAOYSA-N
SMILES: ClC1=C2NC=CC2=CC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results