accessibility menu, dialog, popup

UserName

CAS RN 3590-48-5

ClNHOHNO

CAS RN 3590-48-5

IUPAC Name: 5-(chloromethyl)-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 5-(chloromethyl)-1,3-dihydropyrimidine-2,4-dione
Molecular Weight (g/mol): 160.56
Molecular Formula: C5H5ClN2O2
InChi Key: UCDUBKRXOPMNGH-UHFFFAOYSA-N
SMILES: ClCC1=CNC(=O)NC1=O
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)