CAS RN 35959-50-3
CAS RN 35959-50-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2',5'-Dideoxyuridine, TRC
CAS: 35959-50-3 Molecular Formula: C9H12N2O4 Molecular Weight (g/mol): 212.2 SMILES: C[C@H]1O[C@H](C[C@@H]1O)N2C=CC(=O)NC2=O