accessibility menu, dialog, popup

UserName

CHOClCl

CAS RN 3598-66-1

IUPAC Name: 1,3-dichloro-2-(prop-2-yn-1-yloxy)benzene
Synonyms: 1,3-dichloro-2-(prop-2-yn-1-yloxy)benzene
Molecular Weight (g/mol): 201.05
Molecular Formula: C9H6Cl2O
InChi Key: HCGOBPUSIBTNRV-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(Cl)=C1OCC#C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results