accessibility menu, dialog, popup

UserName

CAS RN 36042-94-1

IUPAC Name: chlorobis(2-methylphenyl)phosphane
Synonyms: chlorobis(2-methylphenyl)phosphane
Molecular Weight (g/mol): 248.69
Molecular Formula: C14H14ClP
InChi Key: KAAGXBGJRWFWPT-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1P(Cl)C1=CC=CC=C1C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results