accessibility menu, dialog, popup

UserName

CH3ONH3CCH3OCH3

CAS RN 3616-56-6

IUPAC Name: (2,2-diethoxyethyl)dimethylamine
Synonyms: (2,2-diethoxyethyl)dimethylamine
Molecular Weight (g/mol): 161.25
Molecular Formula: C8H19NO2
InChi Key: SSFAUOAQOOISRQ-UHFFFAOYSA-N
SMILES: CCOC(CN(C)C)OCC
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results