accessibility menu, dialog, popup

UserName

ClSClN

CAS RN 3622-23-9

IUPAC Name: 2,6-dichloro-1,3-benzothiazole
Synonyms: 2,6-dichloro-1,3-benzothiazole
Molecular Weight (g/mol): 204.07
Molecular Formula: C7H3Cl2NS
InChi Key: QDZGJGWDGLHVNK-UHFFFAOYSA-N
SMILES: ClC1=NC2=CC=C(Cl)C=C2S1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results