accessibility menu, dialog, popup

UserName

H3COOClO

CAS RN 36239-09-5

IUPAC Name: ethyl 3-chloro-3-oxopropanoate
Synonyms: ethyl 3-chloro-3-oxopropanoate
Molecular Weight (g/mol): 150.56
Molecular Formula: C5H7ClO3
InChi Key: KWFADUNOPOSMIJ-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(Cl)=O
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)