CAS RN 362678-52-2
CAS RN 362678-52-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
C3 Ceramide, TRC
CAS: 362678-52-2 Molecular Formula: C21H41NO3 Molecular Weight (g/mol): 355.56 Synonym: N-[(1S,2R,3E)-2-hydroxy-1-(hydroxymethyl)-3-heptadecenyl]propanamide,N-[(1S,2R,3E)-2-hydroxy-1-(hydroxymethyl)-3-heptadecen-1-yl]propanamide IUPAC Name: N-((2S,3R,E)-1,3-dihydroxyoctadec-4-en-2-yl)propionamide SMILES: OC[C@H](NC(CC)=O)[C@H](O)/C=C/CCCCCCCCCCCCC