accessibility menu, dialog, popup

UserName

H2NNHOOCl

CAS RN 36304-40-2

IUPAC Name: 2-(2-chlorophenoxy)acetohydrazide
Synonyms: 2-(2-chlorophenoxy)acetohydrazide
Molecular Weight (g/mol): 200.62
Molecular Formula: C8H9ClN2O2
InChi Key: DSVNKLUHRGOPJC-UHFFFAOYSA-N
SMILES: NNC(=O)COC1=CC=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results