CAS RN 3645-34-9
CAS RN 3645-34-9
IUPAC Name:
titanium(4+) tetrakis(2-ethylhexanoate)
Synonyms:
titanium(4+) tetrakis(2-ethylhexanoate)
Molecular Weight (g/mol):
620.69
Molecular Formula:
C32H60O8Ti
InChi Key:
GDKAXSGMPFSRJY-UHFFFAOYNA-J
SMILES:
[Ti+4].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
1
–
1
of
1
results
1
Titanium(IV) 2-ethylhexanoate, 97%
CAS: 3645-34-9 Molecular Formula: C32H60O8Ti Molecular Weight (g/mol): 620.69 MDL Number: MFCD05663781 InChI Key: GDKAXSGMPFSRJY-UHFFFAOYNA-J Synonym: titanium iv 2-ethylhexanoate,hexanoic acid, 2-ethyl-, titanium 4+ salt,titanium ethylhexanoate,hexanoic acid, 2-ethyl-, titanium 4+ salt 4:1,2-ethylhexanoate; titanium 4+,titanium iv 2-ethylhexanoate 100g,tetrakis 2-ethylhexanoic acid titanium iv salt,titanium 4+ tetrakis 2-ethylhexanoate PubChem CID: 6451578 IUPAC Name: 2-ethylhexanoate;titanium(4+) SMILES: [Ti+4].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O