CAS RN 366-93-8
IUPAC Name:
dihydrogen [(2-chlorophenyl)methyl]({[4-({[(2-chlorophenyl)methyl]amino}methyl)cyclohexyl]methyl})amine dichloride
Synonyms:
dihydrogen [(2-chlorophenyl)methyl]({[4-({[(2-chlorophenyl)methyl]amino}methyl)cyclohexyl]methyl})amine dichloride
Molecular Weight (g/mol):
464.3
Molecular Formula:
C22H30Cl4N2
InChi Key:
NRVIEWRSGDDWHP-UHFFFAOYSA-N
SMILES:
[H+].[H+].[Cl-].[Cl-].ClC1=CC=CC=C1CNCC1CCC(CNCC2=CC=CC=C2Cl)CC1
1
–
2
of
2
results
1
AY 9944 Dihydrochloride, Thermo Scientific Chemicals
CAS: 366-93-8 Molecular Formula: C22H28Cl2N2·2HCl Synonym: 1,4-Bis(2-chlorobenzylaminomethyl)cyclohexane dihydrochloride