accessibility menu, dialog, popup

UserName

OHOO

CAS RN 3663-82-9

IUPAC Name: (2,3-dihydro-1,4-benzodioxin-2-yl)methanol
Synonyms: 2,3-dihydro-1,4-benzodioxin-2-ylmethanol
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: GWQOQQVKVOOHTI-UHFFFAOYNA-N
SMILES: OCC1COC2=CC=CC=C2O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results