accessibility menu, dialog, popup

UserName

(S)(R)(S)OHO(S)(R)OHHOHOOPOOONaNa

CAS RN 3671-99-6

IUPAC Name: disodium (2R,3S,4R,5S)-2,3,4,5-tetrahydroxy-6-(phosphonatooxy)hexanal
Synonyms: disodium (2R,3S,4R,5S)-2,3,4,5-tetrahydroxy-6-(phosphonatooxy)hexanal
Molecular Weight (g/mol): 304.1
Molecular Formula: C6H11Na2O9P
InChi Key: VQLXCAHGUGIEEL-LFGVUSOASA-L
SMILES: [Na+].[Na+].O[C@@H](COP([O-])([O-])=O)[C@@H](O)[C@H](O)[C@@H](O)C=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (2)
  • (1)
  • (9)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (3)