CAS RN 368-16-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
3-N-Methyl-L-histidine, TRC
CAS: 368-16-1 Molecular Formula: C7 H11 N3 O2 Molecular Weight (g/mol): 169.18 Synonym: 3-Methyl-L-histidine,Histidine, 3-methyl-, L- (8CI),3-Methyl-L-histidine,3-Methylhistidine,3-N-Methyl-L-histidine,N3-Methyl-L-histidine IUPAC Name: (2S)-2-amino-3-(3-methylimidazol-4-yl)propanoic acid SMILES: Cn1cncc1C[C@H](N)C(=O)O
2
3-Methyl-L-histidine, MedChemExpress
MedChemExpress 3-Methyl-L-histidine is a biomarker for meat consumption, especially chicken. It is also a biomarker for the consumption of soy products.