accessibility menu, dialog, popup

UserName

CAS RN 36822-11-4

IUPAC Name: 6-phenyl-2-sulfanylidene-1,2,3,4-tetrahydropyrimidin-4-one
Synonyms: 6-phenyl-2-sulfanylidene-1,3-dihydropyrimidin-4-one
Molecular Weight (g/mol): 204.25
Molecular Formula: C10H8N2OS
InChi Key: XEKNACRTWJHOCE-UHFFFAOYSA-N
SMILES: O=C1NC(=S)NC(=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results