CAS RN 36890-68-3
CAS RN 36890-68-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
No Refinements
1
–
0
of
0
results
1
Poly(caprolactone) diol, average M.N. 2000
CAS: 36890-68-3 Molecular Formula: (C6H10O2)n MDL Number: MFCD23713083 SMILES: *-CCCCCC(=O)O-*