accessibility menu, dialog, popup

UserName

H3COFF

CAS RN 369-33-5

IUPAC Name: 1-(3,4-difluorophenyl)ethan-1-one
Synonyms: 1-(3,4-difluorophenyl)ethanone
Molecular Weight (g/mol): 156.13
Molecular Formula: C8H6F2O
InChi Key: VWJSSJFLXRMYNV-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(F)C(F)=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results