accessibility menu, dialog, popup

UserName

CAS RN 3710-84-7

IUPAC Name: N,N-diethylhydroxylamine
Synonyms: diethylhydroxylamine
Molecular Weight (g/mol): 89.14
Molecular Formula: C4H11NO
InChi Key: FVCOIAYSJZGECG-UHFFFAOYSA-N
SMILES: CCN(O)CC
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)