accessibility menu, dialog, popup

UserName

FN

CAS RN 372-48-5

IUPAC Name: 2-fluoropyridine
Synonyms: 2-fluoropyridine
Molecular Weight (g/mol): 97.09
Molecular Formula: C5H4FN
InChi Key: MTAODLNXWYIKSO-UHFFFAOYSA-N
SMILES: FC1=CC=CC=N1
Molecular Weight (g/mol) 
  • (6)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (4)
  • (3)
Percent Purity 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (3)
  • (6)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (3)
  • (4)
  • (3)