accessibility menu, dialog, popup

UserName

CAS RN 37260-88-1

CH3CH3CCHf

CAS RN 37260-88-1

IUPAC Name: hafnium(4+) bis(cyclopenta-1,3-dien-1-ide) dimethanide
Synonyms: hafnium(4+) bis(cyclopenta-1,3-dien-1-ide) dimethanide
Molecular Weight (g/mol): 338.75
Molecular Formula: C12H16Hf
InChi Key: CMPSFTFHQOLFAJ-UHFFFAOYSA-N
SMILES: [CH3-].[CH3-].[Hf+4].C1C=CC=[C-]1.C1C=CC=[C-]1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results