CAS RN 3739-81-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-Methyl-1,2-dihydropyrimidin-2-one, TRC
CAS: 3739-81-9 Molecular Formula: C5H6N2O Molecular Weight (g/mol): 110.12 Synonym: 1-Methyl-2-pyrimidinone,1-Methyl-2-pyrimidone,NSC 193514,1-Methyl-2(1H)-pyrimidinone IUPAC Name: 1-methylpyrimidin-2-one SMILES: CN1C=CC=NC1=O