accessibility menu, dialog, popup

UserName

H3COHFFF

CAS RN 374-01-6

IUPAC Name: 1,1,1-trifluoropropan-2-ol
Synonyms: 1,1,1-trifluoro-2-propanol
Molecular Weight (g/mol): 114.07
Molecular Formula: C3H5F3O
InChi Key: GILIYJDBJZWGBG-UHFFFAOYNA-N
SMILES: CC(O)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)