accessibility menu, dialog, popup

UserName

CAS RN 37408-18-7

CH3ClNCO

CAS RN 37408-18-7

IUPAC Name: 4-chloro-1-isocyanato-2-methylbenzene
Synonyms: 4-chloro-1-isocyanato-2-methylbenzene
Molecular Weight (g/mol): 167.59
Molecular Formula: C8H6ClNO
InChi Key: FTZJLXIATZSKIL-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=CC=C1N=C=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)