accessibility menu, dialog, popup

UserName

H3COHFFFFFFF

CAS RN 375-14-4

IUPAC Name: 3,3,4,4,5,5,5-heptafluoropentan-2-ol
Synonyms: 3,3,4,4,5,5,5-heptafluoropentan-2-ol
Molecular Weight (g/mol): 214.08
Molecular Formula: C5H5F7O
InChi Key: RBPHBIMHZSTIDT-UHFFFAOYNA-N
SMILES: CC(O)C(F)(F)C(F)(F)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results