accessibility menu, dialog, popup

UserName

CAS RN 37593-03-6

H3CCH3O

CAS RN 37593-03-6

IUPAC Name: 1-(4-heptylphenyl)ethan-1-one
Synonyms: 1-(4-heptylphenyl)ethanone
Molecular Weight (g/mol): 218.34
Molecular Formula: C15H22O
InChi Key: UQBRZOXCKKBKDU-UHFFFAOYSA-N
SMILES: CCCCCCCC1=CC=C(C=C1)C(C)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results