accessibility menu, dialog, popup

UserName

HOONHN

CAS RN 37718-11-9

IUPAC Name: 1H-pyrazole-4-carboxylic acid
Synonyms: 1H-pyrazole-4-carboxylic acid
Molecular Weight (g/mol): 112.09
Molecular Formula: C4H4N2O2
InChi Key: IMBBXSASDSZJSX-UHFFFAOYSA-N
SMILES: OC(=O)C1=CNN=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)