accessibility menu, dialog, popup

UserName

BrBr

CAS RN 37763-43-2

IUPAC Name: 1-bromo-2-(bromomethyl)naphthalene
Synonyms: 1-bromo-2-(bromomethyl)naphthalene
Molecular Weight (g/mol): 299.99
Molecular Formula: C11H8Br2
InChi Key: DQTOCXIHYIQHCK-UHFFFAOYSA-N
SMILES: BrCC1=CC=C2C=CC=CC2=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results