accessibility menu, dialog, popup

UserName

ClSN

CAS RN 37859-43-1

IUPAC Name: 2-(chloromethyl)-1,3-benzothiazole
Synonyms: 2-(chloromethyl)-1,3-benzothiazole
Molecular Weight (g/mol): 183.65
Molecular Formula: C8H6ClNS
InChi Key: SERUZNHRWBXDOX-UHFFFAOYSA-N
SMILES: ClCC1=NC2=CC=CC=C2S1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)