accessibility menu, dialog, popup

UserName

CH3OOSSH3CNH2

CAS RN 379247-26-4

IUPAC Name: ethyl 5'-amino-5-methyl-[2,3'-bithiophene]-4'-carboxylate
Synonyms: ethyl 5'-amino-5-methyl-[2,3'-bithiophene]-4'-carboxylate
Molecular Weight (g/mol): 267.36
Molecular Formula: C12H13NO2S2
InChi Key: OCXFDSDTIGSDOV-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)SC=C1C1=CC=C(C)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results