accessibility menu, dialog, popup

UserName

FFFN

CAS RN 3796-24-5

IUPAC Name: 4-(trifluoromethyl)pyridine
Synonyms: 4-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 147.1
Molecular Formula: C6H4F3N
InChi Key: IIYVNMXPYWIJBL-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=NC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results