accessibility menu, dialog, popup

UserName

BOOCH3HOOH(E/Z)

CAS RN 380430-58-0

IUPAC Name: [4-(3-methoxy-3-oxoprop-1-en-1-yl)phenyl]boronic acid
Synonyms: 4-(3-methoxy-3-oxoprop-1-en-1-yl)phenylboronic acid
Molecular Weight (g/mol): 206
Molecular Formula: C10H11BO4
InChi Key: BIKAXBPCHWDATP-UHFFFAOYSA-N
SMILES: COC(=O)C=CC1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results