CAS RN 380844-49-5
CAS RN 380844-49-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-Chloro-1-desmethylpiperazinyl-bosutinib, TRC
CAS: 380844-49-5 Molecular Formula: C21H18Cl3N3O3 Molecular Weight (g/mol): 466.74 Synonym: 7-(3-Chloropropoxy)-4-((2,4-dichloro-5-methoxyphenyl)amino)-6-methoxyquinoline-3-carbonitrile IUPAC Name: 7-(3-chloropropoxy)-4-(2,4-dichloro-5-methoxyanilino)-6-methoxyquinoline-3-carbonitrile SMILES: ClCCCOC(C(OC)=C1)=CC2=C1C(NC3=CC(OC)=C(Cl)C=C3Cl)=C(C#N)C=N2