accessibility menu, dialog, popup

UserName

OHNBrI

CAS RN 381233-75-6

IUPAC Name: 5-bromo-3-iodo-1,2-dihydropyridin-2-one
Synonyms: 5-bromo-3-iodo-1H-pyridin-2-one
Molecular Weight (g/mol): 299.89
Molecular Formula: C5H3BrINO
InChi Key: WAUPPDAXWKLNNU-UHFFFAOYSA-N
SMILES: BrC1=CNC(=O)C(I)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results