accessibility menu, dialog, popup

UserName

OOOOOOOOONdNd

CAS RN 38245-38-4

IUPAC Name: dineodymium(3+) hydrate tricarbonate
Synonyms: dineodymium(3+) hydrate tricarbonate
Molecular Weight (g/mol): 486.52
Molecular Formula: C3H2Nd2O10
InChi Key: CZXBEKQIOBCHSG-UHFFFAOYSA-H
SMILES: O.[Nd+3].[Nd+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results