accessibility menu, dialog, popup

UserName

CH3SiH3CCH3Br

CAS RN 38274-16-7

IUPAC Name: [2-(2-bromophenyl)ethynyl]trimethylsilane
Synonyms: [2-(2-bromophenyl)ethynyl]trimethylsilane
Molecular Weight (g/mol): 253.21
Molecular Formula: C11H13BrSi
InChi Key: FABNGXSCLHXUOH-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C#CC1=CC=CC=C1Br
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results