accessibility menu, dialog, popup

UserName

ClNNN

CAS RN 3842-55-5

IUPAC Name: 2-chloro-4,6-diphenyl-1,3,5-triazine
Synonyms: 2-chloro-4,6-diphenyl-1,3,5-triazine
Molecular Weight (g/mol): 267.72
Molecular Formula: C15H10ClN3
InChi Key: DDGPPAMADXTGTN-UHFFFAOYSA-N
SMILES: ClC1=NC(=NC(=N1)C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results