accessibility menu, dialog, popup

UserName

H3CH3CCH3OONHN

CAS RN 38427-94-0

IUPAC Name: tert-butyl N-(pyridin-2-yl)carbamate
Synonyms: tert-butyl N-(pyridin-2-yl)carbamate
Molecular Weight (g/mol): 194.23
Molecular Formula: C10H14N2O2
InChi Key: ORUGTGTZBRUQIT-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=CC=CC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results