accessibility menu, dialog, popup

UserName

ONOFClF

CAS RN 3847-58-3

IUPAC Name: 2-chloro-1,3-difluoro-4-nitrobenzene
Synonyms: 2-chloro-1,3-difluoro-4-nitrobenzene
Molecular Weight (g/mol): 193.53
Molecular Formula: C6H2ClF2NO2
InChi Key: CTVBVNKEMCGZDF-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(F)C(Cl)=C1F
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)