accessibility menu, dialog, popup

UserName

H3C(S)NH3CH3CH3H3C

CAS RN 3850-30-4

IUPAC Name: (2S)-3,3-dimethylbutan-2-aminium
Synonyms: (2S)-3,3-dimethylbutan-2-aminium
Molecular Weight (g/mol): 102.2
Molecular Formula: C6H16N
InChi Key: DXSUORGKJZADET-YFKPBYRVSA-O
SMILES: C[C@H]([NH3+])C(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results