CAS RN 3869-32-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6Beta-Hydroxy Triamcinolone Acetonide, TRC
CAS: 3869-32-7 Molecular Formula: C24 H31 F O7 Molecular Weight (g/mol): 450.5 Synonym: (6β,11β,16α)-9-Fluoro-6,11,21-trihydroxy-16,17-[(1-methylethylidene)bis(oxy)]-pregna-1,4-diene-3,20-dione; SMILES: CC1(C)O[C@@H]2C[C@H]3[C@@H]4C[C@@H](O)C5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO