accessibility menu, dialog, popup

UserName

CH3ONH2

CAS RN 388072-09-1

IUPAC Name: 1-(3-methylfuran-2-yl)methanamine
Synonyms: 1-(3-methylfuran-2-yl)methanamine
Molecular Weight (g/mol): 111.14
Molecular Formula: C6H9NO
InChi Key: CTGIVQJZUKJDNH-UHFFFAOYSA-N
SMILES: CC1=C(CN)OC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results