accessibility menu, dialog, popup

UserName

CH3OClO

CAS RN 38870-89-2

IUPAC Name: 2-methoxyacetyl chloride
Synonyms: methoxy-acetyl chloride
Molecular Weight (g/mol): 108.52
Molecular Formula: C3H5ClO2
InChi Key: JJKWHOSQTYYFAE-UHFFFAOYSA-N
SMILES: COCC(Cl)=O
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)
  • (3)